Learn More
Medchemexpress LLC Fmoc-Asn(Trt)-OH | 132388-59-1 | 596.67 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002327
Encompass_Preferred
Fmoc-Asn(Trt)-OH is an Fmoc-protected asparagine derivative that supports solid-phase peptide synthesis. It is intended for research use only.
  • Fmoc-protected asparagine derivative
  • Supports solid-phase peptide synthesis
  • Trityl-protected side-chain carboxamide
  • Trityl group stabilizer
  • Useful in biochemical assays
  • Acts as an inhibitor
Catalog No. 50-004-49410