Learn More
Medchemexpress LLC Fmoc-Asn(Trt)-OH-13C4,15N2 | 1217456-18-2 | 602.63 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW002327S1MG
Encompass_Preferred
Fmoc-Asn(Trt)-OH-13C4,15N2 is a 15N-labeled and 13C-labeled Fmoc-Asn(Trt)-OH. This biochemical reagent can be used as a biological material or organic compound for life science related research and for the synthesis of Fmoc-based solid-phase peptide synthesis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-48961