Learn More
Apexbio Technology LLC Fmoc-Asn(Trt)-OH 132388-59-1 25g
SDP
Supplier:  Apexbio Technology LLC A735725000
Encompass_Preferred
Fmoc-Asn Trt -OH (CAS 132388-59-1) is an amino acid derivative used as a biochemical reagent in peptide synthesis It is designed for Fmoc-based solid-phase peptide synthesis incorporating an N-terminal Fmoc protecting group and a side-chain trityl (Trt) group on asparagine to ensure selective protection and facilitate peptide assembly Researchers employ Fmoc-Asn Trt -OH extensively for synthesizing peptides or protein fragments to enable structural studies functional assays mechanistic investigations of biological processes and preliminary pharmaceutical research
Catalog No. 50-224-0853