Learn More
Medchemexpress LLC Fmoc-Asp-NH2 | 200335-40-6 | MFCD00151920 | 98.5% | 354.36 | C19H18N2O5 | 1 ML

Supplier: Medchemexpress LLC HY13541810MM 1ML
Fmoc-Asp-NH2 is a cleavable antibody-drug conjugate (ADC) linker provided as a ready-to-use solution or as a solid for reconstitution, intended for research use.
- 10 mM, 1 mL solution in DMSO.
- Cleavable ADC linker for synthesis of antibody-drug conjugates and related conjugation chemistries.
- Purity 98.5% (typical).
- Molecular formula C19H18N2O5; molecular weight 354.36 g/mol.
- CAS number 200335-40-6.
- Store in solvent at -80°C (6 months) or -20°C (1 month); powder storage varies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.