Learn More
Medchemexpress LLC Fmoc-Asp(OAll)-OH | 146982-24-3 | 395.41 | 50 G

Supplier: Medchemexpress LLC HYW00807550G
Fmoc-Asp(OAll)-OH is an aspartic acid derivative recognized for its beneficial properties as an ergogenic dietary substance. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, providing fuel during exercise, aiding mental performance during stress-related tasks, and helping prevent exercise-induced muscle damage.
- Purity: 99.89%
- Molecular weight: 395.41
- Formula: C22H21NO6
- CAS number: 146982-24-3
- Appearance: Solid
- Color: White to off-white
- Solubility: Soluble in DMSO at 100 mg/mL (252.90 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.