Learn More
MilliporeSigma FMOC-ASP(ODMAB)-OH
Supplier:  MilliporeSigma 8520780001
Encompass
Fmoc-Asp(ODmab)-OH; > /=95% by HPLC; Solid; EMD; Novabiochem; 1g; N-alpha-Fmoc-L-aspartic acid Beta-4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-amino}benzyl ester; F.W. 666.7; C39H42N
Catalog No. 50-230-1868