Learn More
Medchemexpress LLC Fmoc-asp-ome | 145038-52-4 | MFCD03701473 | 99.3% | 369.37 g/mol | C20H19NO6 | 500 G

Supplier: Medchemexpress LLC HYW041982500G
Fmoc-Asp-OMe is the 9-fluorenylmethoxycarbonyl-protected methyl ester of aspartic acid, used as an Fmoc-protected amino acid building block in solid-phase peptide synthesis and organic synthesis applications.
- used as an Fmoc-protected aspartic acid building block for peptide synthesis
- chemical formula: C20H19NO6
- molecular weight: 369.37 g/mol
- purity typically 99.3%
- appearance: white to off-white solid
- solubility: soluble in DMSO (100 mg/mL) and formulatable for in vivo use with common co-solvents
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.