Learn More
Apexbio Technology LLC Fmoc-Asp-OtBu 129460-09-9 25g
SDP
Supplier:  Apexbio Technology LLC A736425000
Encompass_Preferred
Fmoc-Asp-OtBu (CAS 129460-09-9) is an Fmoc-protected aspartic acid derivative with a tert-butyl ester (OtBu) protecting group on its side-chain carboxyl functionality It is specifically employed in solid-phase peptide synthesis protocols where it facilitates controlled stepwise assembly of peptide chains by allowing selective deprotection of the Fmoc group under basic conditions and removal of the OtBu group under acidic conditions Based on these properties Fmoc-Asp-OtBu holds research potential in the synthesis of peptides containing specific aspartic acid residues thereby supporting structural and functional studies of peptide sequences and interactions in biomolecular research
Catalog No. 50-000-97137