Learn More
Bachem Fmoc-cis-4-fluoro-Pro-OH
SDP
Supplier:  Bachem 4034298.5000
Encompass_Preferred
Fmoc-cis-4-fluoro-Pro-OH, 203866-19-7, C20H18FNO4, Mr 355.37, 5g. Fmoc-(2S,4S)-4-fluoro-pyrrolidine-2-carboxylic acid, Fmoc-flp-OH. Stabilizes the PPII helix in Pro-rich short peptides, though less efficiently than the trans isomer. Oligomers of flp were studied by Horng and Raines.
Catalog No. 50-258-818