Learn More
Medchemexpress LLC Fmoc-Cit-PAB-OH | 870487-04-0 | MFCD32062910 | >97.0% | 502.56 | C28H30N4O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1417655G
Encompass_Preferred
Fmoc-Cit-PAB-OH is an Fmoc-protected peptide linker containing a citrulline residue and a para-aminobenzyl (PAB) spacer. The Fmoc protecting group is removable under basic conditions to reveal a free amine for downstream conjugation chemistry. Supplied as a powder, the reagent is intended for linker synthesis and conjugation workflows and includes manufacturer guidance on storage and solubility.
  • Fmoc-protected amine enables orthogonal deprotection.
  • Citrulline residue provides a peptide handle for conjugation.
  • Para-aminobenzyl spacer facilitates drug-linker release mechanisms.
  • High purity (>97.0% HPLC) suitable for research applications.
  • Powder form with defined cold-storage conditions.
  • Soluble in DMSO at 70 mg/mL; may require ultrasonication.
Catalog No. 50-002-83564