Learn More
MilliporeSigma FMOC-CYS(ACM)-OH
Supplier:  MilliporeSigma 8520060005
Encompass
Fmoc-Cys(Acm)-OH; > /=97% by HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-S-acetamidomethyl-L-cysteine; F.W. 414.5; C21H22N2O5S; M.P. 147-150 deg.C (296.6-302 deg.F); CAS: 86060-
Catalog No. 50-230-1593