Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Cys(Acm)-OH | 86060-81-3 | 99.77% | 414.47 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW01314310G

Encompass_Preferred

Fmoc-Cys(Acm)-OH is a cysteine derivative. Amino acids and their derivatives are utilized as ergogenic supplements due to their impact on anabolic hormone secretion, their role as a fuel source during exercise, their ability to enhance mental performance during stressful tasks, and their capacity to prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Purity: 99.77%
  • Molecular weight: 414.47
  • Formula: C21H22N2O5S
  • CAS No.: 86060-81-3
  • Appearance: Solid, white to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility (In vitro): 100 mg/mL in DMSO (241.27 mM); requires sonication; hygroscopic DMSO can significantly affect solubility, so newly opened DMSO should be used.

Catalog No. 50-004-54580


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.