Learn More
Medchemexpress LLC Fmoc-Cys-Asp10 TFA | 2407782-70-9 | 1494.27 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP3497A1MG
Encompass_Preferred
Fmoc-Cys-Asp10 is a non-releasable oligopeptide linker involved in the synthesis of releasable oligopeptide linkers. Releasable oligopeptide linkers can be used to deliver agents to a bone fracture-homing oligopeptide, which may reduce fractured femur healing times.
  • Involved in the synthesis of releasable oligopeptide linkers
  • Can be used to deliver an agent to a bone fracture-homing oligopeptide
  • Reduces fractured femur healing times
Catalog No. 50-003-98291