Learn More
Bachem Fmoc-Cys(NPys)-OH
SDP
Supplier:  Bachem 4016999.0001
Encompass_Preferred
Fmoc-Cys(NPys)-OH, 159700-51-3, C23H19N3O6S2, Mr 497.55, 1g. Npys allows site-directed disulfide bond formation, as it is cleaved by thiols.
Catalog No. NC1402669