Learn More
Medchemexpress LLC Fmoc-Cys-OtBu 2 di 5g
SDP
Supplier:  Medchemexpress LLC HYW1019545G
Encompass_Preferred
(Fmoc-Cys-OtBu)2 (Disulfide bond) is a cystine dipeptide which is linked through an oxidized disulfide bond (Fmoc-Cys-OtBu)2 (disulfide bridge) can be used in the synthesis of cleavable disulfide linkers
Catalog No. 50-002-05013