Learn More
Bachem Fmoc-Cys(SO3H)-OH · disodium salt
SDP
Supplier:  Bachem 4016589.0001
Encompass_Preferred
Fmoc-Cys(SO3H)-OH · disodium salt, 163558-30-3, C18H15NNa2O7S2, Mr 467.43, 1g. A useful cysteine derivative for solid-phase synthesis by the Fmoc-strategy. The S-sulfonate group is stable towards TFA and is cleaved either with phosphines or with mercaptans.
Catalog No. 50-312-787