Learn More
Bachem Fmoc-Cys(Trt)-OH
SDP
Supplier:  Bachem 4007272.0005
Encompass_Preferred
Fmoc-Cys(Trt)-OH, 103213-32-7, C37H31NO4S, Mr 585.72, 5g. Standard derivative for Fmoc-SPPS. Educt for obtaining S-alkylated cysteines via thiol-ene reaction.
Catalog No. 50-312-647