Learn More
Apexbio Technology LLC Fmoc-Cys(Trt)-ol 215954-72-6 25g
SDP
Supplier:  Apexbio Technology LLC A660025000
Encompass_Preferred
Fmoc-Cys Trt -ol is a protected cysteine derivative designed for solid-phase peptide synthesis (SPPS) It incorporates an N-terminal Fmoc group and a trityl (Trt) protecting group on the cysteine thiol side-chain preventing oxidation or disulfide bridge formation during synthesis Fmoc-Cys Trt -ol serves as a building block for the precise incorporation of cysteine residues into peptides or protein fragments without compromising sulfhydryl functionality Based on these properties Fmoc-Cys Trt -ol holds research potential in the synthesis of sulfhydryl-functionalized peptides bioconjugates and peptide-drug conjugates for biomedical research and therapeutic development
Catalog No. 50-000-93564