Learn More
STA PHARMACEUTICAL US LLC Fmoc-D-1,2,3,4-Tetrahydronorharman-3-carboxylic acid | 1 g | CAS 268731-07-3 | MDL MFCD01861321
Supplier: STA PHARMACEUTICAL US LLC STAAA109541GM
Fmoc-D-1,2,3,4-Tetrahydronorharman-3-carboxylic acid is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 268731-07-3
- MDL: MFCD01861321
- InChIKey: IHHULIKSMJSELI-RUZDIDTESA-N
- Molecular Weight: 438.483
- Molecular Formula: C27H22N2O4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (R)-2-(((9H-fluoren-9-yl)methoxy)carbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid
- SMILES: O=C(N1CC2=C(C3=CC=CC=C3N2)C[C@@H]1C(O)=O)OCC4C5=CC=CC=C5C6=CC=CC=C64
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.