Learn More
Apexbio Technology LLC Fmoc-D-2-Me-Trp(Synonyms: Fmoc-D-2-Methyltryptophan, Fmoc-D-2-Me-Trp-OH, N-alpha-Fmoc-D-2-Me-Trp, Fmoc-D-α-Methyltryptophan), 1g, CAS: 196808-79-4.
SDP
Supplier:  Apexbio Technology LLC A66941000
Encompass_Preferred
Fmoc-D-2-Me-Trp (CAS 196808-79-4) is an Fmoc-protected D-form methylated tryptophan derivative frequently utilized as an amino acid building block in peptide synthesis and structural modifications Its methyl substitution at the side chain serves to modulate peptide conformation thereby influencing binding interactions between peptide ligands and protein receptors By altering ligand-protein interface specificity and stability this compound facilitates investigation into protein-ligand recognition mechanisms making it valuable in peptide-based pharmacological and medicinal chemistry research
Catalog No. 50-000-60542