Learn More
Medchemexpress LLC Fmoc-D-3,3-diphenylalanine | 189937-46-0 | MFCD00672338 | 99.4% | 463.52 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0089221G
Encompass_Preferred
Fmoc-D-3,3-diphenylalanine is a peptide derivative used for research purposes. It is available in solid, white to off-white appearance.
  • High purity (99.43% with ee.: 99.08%)
  • Available in various quantities (1g, 5g, 10g, 50g)
  • Detailed documentation including Data Sheet, COA, SDS, HNMR, RP-HPLC, and Handling Instructions
  • Good solubility in DMSO (200 mg/mL)
  • Clear storage recommendations for powder and in-solvent forms
  • Tools available for calculating molarity, dilution, and in vivo dissolution
Catalog No. 50-004-51207