Learn More
MilliporeSigma FMOC-D-ARG(PBF)-OH
Supplier:  MilliporeSigma 8521650005
Encompass
Fmoc-D-Arg(Pbf)-OH; > /=97% by HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-N-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-D-arginine; F.W. 648.8; C34H40N4O7S; CAS: 1876
Catalog No. 50-230-2904