Learn More
MilliporeSigma FMOC-D-ASP(OTBU)-OH
Supplier:  MilliporeSigma 8521540005
Encompass
Fmoc-D-Asp(OtBu)-OH; > /=98% by HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-D-aspartic acid beta-tert-butyl ester; F.W. 411.5; C23H25NO6; CAS: 112883-39-3
Catalog No. 50-230-2863