Learn More
STA PHARMACEUTICAL US LLC Fmoc-D-beta-Homo-hGlu(OtBu)-OH | 10 g | CAS 1421258-67-4 | MDL MFCD22887410
Supplier: STA PHARMACEUTICAL US LLC STAAA1176110GM
Fmoc-D-beta-Homo-hGlu(OtBu)-OH is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1421258-67-4
- MDL: MFCD22887410
- InChIKey: XPCDWOCHPTYDPV-MRXNPFEDSA-N
- Molecular Weight: 439.508
- Molecular Formula: C25H29NO6
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(tert-butoxy)-6-oxohexanoic acid
- SMILES: CC(C)(OC(CC[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC(O)=O)=O)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.