Learn More
Medchemexpress LLC Fmoc-D-Dab(Boc)-OH | 114360-56-4 | MFCD00798647 | 99.1% | 440.49 g/mol | C24H28N2O6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0190321G
Encompass_Preferred
Fmoc-D-Dab(Boc)-OH is an Fmoc-protected D-2,4-diaminobutyric acid derivative for incorporation into peptides via solid-phase peptide synthesis. It features an Fmoc-protected N-terminus and a Boc-protected side-chain amine to enable orthogonal deprotection and selective coupling. Supplied as a high-purity solid, it is suitable for research synthesis and structure-activity studies, including antibacterial peptide design.
  • Fmoc-protected N-terminus for standard Fmoc-SPPS workflows.
  • Boc-protected side-chain amine for orthogonal deprotection.
  • High purity suitable for research synthesis (99.1%).
  • Solid, white to off-white appearance for easy handling.
  • Stable storage at room temperature with extended shelf life.
  • Enables incorporation of D-diaminobutyric acid residues for SAR studies.
Catalog No. 50-003-00840