Learn More
Medchemexpress LLC Fmoc-D-Dap(Boc)-OH | 198544-42-2 | MFCD00798648 | 99.6% | 426.46 | C23H26N2O6 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW04635550G
Encompass_Preferred
Fmoc-D-Dap(Boc)-OH is an Fmoc-protected D-2,3-diaminopropionic acid derivative used as an amino acid building block for solid-phase peptide synthesis and the preparation of peptide mimetics. The product is supplied for research use only.
  • Used as an amino acid building block for solid-phase peptide synthesis.
  • Contains Fmoc and Boc protecting groups for orthogonal deprotection.
  • Suitable for preparation of DOTA-modified peptides and metal chelates.
  • White to off-white solid with high purity (about 99.6%).
Catalog No. 50-002-10050