Learn More
Medchemexpress LLC Fmoc-d-dap(boc)-oh | 198544-42-2 | MFCD00798648 | 99.6% | 426.46 g/mol | C23H26N2O6 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW046355
Encompass_Preferred
Fmoc-D-Dap(Boc)-OH is an Fmoc-protected derivative of D-diaminopropionic acid with a Boc-protected side-chain amino group. Supplied as a white to off-white solid for use as a building block in peptide synthesis and in preparation of peptide-based chelators and related metal complexes for research applications.
  • Fmoc-protected α-amino group
  • Boc-protected side-chain amino group
  • Suitable for solid-phase peptide synthesis
  • High purity (99.6%)
  • Supplied as a 25 G solid
  • Storage stable at room temperature for up to 3 years
Catalog No. 50-002-08843