Learn More
STA PHARMACEUTICAL US LLC Fmoc-D-norArg(Z)2-OH | 5 g | CAS 2044710-16-7 | MDL MFCD15142018
Supplier: STA PHARMACEUTICAL US LLC STAAA121425GM
Fmoc-D-norArg(Z)2-OH is a Amino Acid reagent (Subcategory: Arg) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 2044710-16-7
- MDL: MFCD15142018
- InChIKey: OJBZBKXVLYDJEI-HKBQPEDESA-N
- Molecular Weight: 650.688
- Molecular Formula: C36H34N4O8
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29252900
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-((3,7-dioxo-1,9-diphenyl-2,8-dioxa-4,6-diazanonan-5-ylidene)amino)butanoic acid
- SMILES: O=C(OCC1=CC=CC=C1)N/C(NC(OCC2=CC=CC=C2)=O)=N/CC[C@H](C(O)=O)NC(OCC3C4=CC=CC=C4C5=CC=CC=C53)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.