Learn More
Medchemexpress LLC Fmoc-D-Phe(4-CF3)-OH | 238742-88-6 | 455.43 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW013123
Encompass_Preferred
Fmoc-D-Phe(4-CF3)-OH is a Phenylalanine derivative that can be used for the research of peptide inhibitors of protein-protein interactions.
  • Phenylalanine derivative
  • Used for research of peptide inhibitors of protein-protein interactions
  • Stable as powder for up to 3 years at -20°C
  • Stable in solvent for up to 6 months at -80°C
  • Soluble in DMSO
Catalog No. 50-003-67049