Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-D-Phe(4-CN)-OH | 205526-34-7 | 98.0% | 412.44 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0109575G

Encompass_Preferred

Fmoc-D-Phe(4-CN)-OH is a derivative of phenylalanine. Amino acids and their derivatives are utilized commercially as ergogenic supplements. They are known to influence the secretion of anabolic hormones, provide fuel during exercise, enhance mental performance during stress-related tasks, and help prevent exercise-induced muscle damage. These substances are recognized for their beneficial role as ergogenic dietary components.

  • Molecular weight: 412.44
  • Formula: C25H20N2O4
  • CAS No.: 205526-34-7
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-66319


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.