Learn More
Medchemexpress LLC Fmoc-D-Pra-OH | 220497-98-3 | 335.36 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00839510G
Encompass_Preferred
Fmoc-D-Pra-OH is a Glycine derivative and a click chemistry reagent. It contains an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. Amino acids and amino acid derivatives, such as Fmoc-D-Pra-OH, are utilized as ergogenic supplements that influence anabolic hormone secretion, supply fuel during exercise, improve mental performance during stress, and prevent exercise-induced muscle damage. These compounds are recognized as beneficial ergogenic dietary substances.
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Influences anabolic hormone secretion
  • Provides fuel during exercise
  • Improves mental performance during stress
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-47292