Learn More
MilliporeSigma FMOC-D-PRO-OH
Supplier:  MilliporeSigma 8521490025
Encompass
Fmoc-D-Pro-OH; > /=98%; Solid; EMD; Novabiochem; 25g; N-alpha-Fmoc-D-proline; F.W. 337.4; C20H19NO4; CAS: 101555-62-8;
Catalog No. 50-230-2832