Learn More
Medchemexpress LLC Fmoc-D-Pro-OH | 101555-62-8 | 99.9% | 337.37 | 100 G

Supplier: Medchemexpress LLC HYW009124100G
Fmoc-D-Pro-OH is a proline derivative. Amino acids and amino acid derivatives are utilized commercially as ergogenic supplements. They influence the secretion of anabolic hormones, contribute to fuel supply during exercise, enhance mental performance during stressful tasks, and help prevent exercise-induced muscle damage. These compounds are recognized for their beneficial properties as ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Store at room temperature for 3 years
- Solubility: In Vitro: DMSO: 100 mg/mL (296.41 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO)
- Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.