Learn More
Medchemexpress LLC Fmoc-D-α-t-butylglycine | 198543-64-5 | MFCD00065650 | 98.0% | 353.41 g/mol | C21H23NO4 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW04871850G
Encompass_Preferred
Fmoc-D-Tle-OH is an Fmoc-protected D-amino acid used as a building block in peptide synthesis and small-molecule assembly. It introduces D-tert-leucine residues and is used to prepare chelating agents that can form stereoisomer-enriched metal complexes.
  • Fmoc-protected D-tert-leucine building block.
  • Suitable for peptide synthesis and solid-phase peptide synthesis.
  • Useful for preparing stereochemically defined chelating agents.
  • Supplied as a powdered solid for convenient weighing and handling.
  • Typically high purity for research applications.
Catalog No. 50-002-09901