Learn More
Medchemexpress LLC Fmoc-D-Thi-OH | 201532-42-5 | 99.1% | 393.46 g/mol | C22H19NO4S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0487221G
Encompass_Preferred
Fmoc-D-Thi-OH is an Fmoc-protected D-amino acid used as a building block in peptide synthesis and small-molecule assembly. The Fmoc protecting group permits standard Fmoc solid-phase peptide synthesis workflows, and the thienyl side chain enables incorporation of a thienylalanine residue for medicinal chemistry applications.
  • Suitable for Fmoc solid-phase peptide synthesis.
  • Contains a D-2-thienylalanine side chain for structural variation in peptides.
  • Provides a stable Fmoc protecting group compatible with standard deprotection conditions.
  • Available in multiple pack sizes for small-scale synthesis and method development.
  • High chemical purity for reliable incorporation into peptide sequences.
Catalog No. 50-002-10114