Learn More
Medchemexpress LLC Fmoc-D-Tyr(Me)-OH | 201335-88-8 | MFCD00237393 | 99.6% | 417.45 g/mol | C25H23NO5 | 5 G

Supplier: Medchemexpress LLC HYW0222275G
Fmoc-D-Tyr(Me)-OH is an Fmoc-protected O-methyl derivative of D-tyrosine used as a protected amino acid building block for peptide synthesis and related research. Supplied as a solid and as ready-to-use DMSO solutions for convenient handling and incorporation into synthetic workflows.
- Fmoc-protected O-methyl D-tyrosine for peptide synthesis.
- CAS: 201335-88-8.
- Chemical formula C25H23NO5.
- Molecular weight 417.45 g/mol.
- High purity (≈99.6%).
- Available as bulk powder and 10 mM DMSO solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.