Learn More
Medchemexpress LLC Fmoc-D-Tyr(tBu)-OH | 118488-18-9 | 459.53 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008426
Encompass_Preferred
Fmoc-D-Tyr(tBu)-OH is a tyrosine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, and mental performance under stress. They also help prevent exercise-induced muscle damage and are recognized as beneficial ergogenic dietary substances. This product is a white to off-white solid with an enantiomeric excess of 100.00% and its ¹H NMR spectrum is consistent with its structure.
  • Tyrosine derivative
  • Used as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Aids fuel supply during exercise
  • Improves mental performance during stress
  • Prevents exercise-induced muscle damage
  • White to off-white solid
  • Enantiomeric excess of 100.00%
Catalog No. 50-004-49912