Learn More
Medchemexpress LLC Fmoc-Dab(Boc)-OH | 125238-99-5 | 440.49 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0080245G
Encompass_Preferred
Fmoc-Dab(Boc)-OH is an amino acid derivative with an Fmoc protecting group. This compound can be used to synthesize somatostatin analogs that inhibit neointima formation induced by balloon injury in rats without altering growth hormone release. It is also suitable for the general synthesis of peptides.
  • Amino acid derivative with Fmoc protecting group
  • Can be used to synthesize somatostatin analogs
  • Inhibits neointima formation induced by balloon injury
  • Suitable for general peptide synthesis
  • Available as an off-white to light yellow solid
Catalog No. 50-004-46182