Learn More
Medchemexpress LLC Fmoc-Dap(Boc)-OH | 162558-25-0 | 100.0% | 426.46 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01092210G
Encompass_Preferred
Fmoc-Dap(Boc)-OH is an amino acid derivative featuring an Fmoc protecting group. It is utilized in the synthesis of bicyclic peptide tachykinin NK2 antagonists. This product is intended for research use only and is not sold to patients.
  • Amino acid derivative
  • Contains an Fmoc protecting group
  • Used to synthesize bicyclic peptide tachykinin NK2 antagonists
  • For research purposes only
Catalog No. 50-004-51984