Learn More
Apexbio Technology LLC Fmoc-Dap(Boc)-OH 162558-25-0 5g
SDP
Supplier:  Apexbio Technology LLC A74985000
Encompass_Preferred
Fmoc-Dap Boc -OH is an amino acid derivative routinely used in solid-phase peptide synthesis to facilitate the introduction of diaminopropionic acid (Dap) units into peptide sequences The Fmoc group can be removed under standard deprotection conditions to expose a free amino functionality allowing subsequent peptide coupling Fmoc-Dap Boc -OH is particularly suitable for constructing cyclic and bicyclic peptides as well as peptide analogs targeting neurokinin NK2 receptor systems Based on these properties Fmoc-Dap Boc -OH holds research potential in pharmacological investigations and molecular studies involving receptor binding signaling pathways and receptor-ligand interaction analyses
Catalog No. 50-000-93247