Learn More
Bachem Fmoc-e-aminocaproic acid
SDP
Supplier:  Bachem 4006429.0025
Encompass_Preferred
Fmoc-e-aminocaproic acid, 88574-06-5, C21H23NO4, Mr 353.42, 25g. Fmoc-eAhx-OH, Fmoc-6-aminohexanoic acid, Fmoc-LC-OH. Protected flexible non-polar spacer moiety also abbreviated Fmoc-e-Aca, Fmoc-e-Ahx, or Fmoc-LC for use in Fmoc-SPPS. Bachem additionally offers Fmoc-eAhx-Wang resin (D-1050).
Catalog No. 50-258-552