Learn More
Bachem Fmoc-amino-PEG2-ethylamino-Suc-OH
SDP
Supplier:  Bachem 4082329.0025
Encompass_Preferred
Fmoc-amino-PEG2-ethylamino-Suc-OH, 613245-91-3, C25H30N2O7, Mr 470.52, 25g. N-(Fmoc-amino-PEG2-ethyl)-succinamic acid, N-(Fmoc-8-amino-3,6-dioxa-octyl)-succinamic acid, Fmoc-Ebes-OH. Fmoc-Ebes, short PEG spacer, which can be incorporated into peptides during SPPS.
Catalog No. 50-194-7393