Learn More
Medchemexpress LLC Fmoc-GGFG-DXd | 1599440-11-5 | 99.8% | 1063.09 g/mol | C57H55FN8O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15684825MG
Encompass_Preferred
Fmoc-GGFG-DXd is a drug-linker conjugate used as a building block in antibody-drug conjugate (ADC) research and synthesis.
  • Drug-linker conjugate for ADC synthesis.
  • Contains Fmoc-protected GGFG peptide linker and DXd topoisomerase I inhibitor.
  • Available in a 25 mg research pack.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-04437