Learn More
Apexbio Technology LLC Fmoc-Gln(Trt)-OH 132327-80-1 100g
SDP
Supplier:  Apexbio Technology LLC A7881100000
Encompass_Preferred
Fmoc-Gln Trt -OH (132327-80-1) is a chemically protected amino acid derivative of glutamine It is primarily employed as a monomer in solid-phase peptide synthesis (SPPS) The compound features a 9-fluorenylmethoxycarbonyl (Fmoc) group protecting the alpha-amino position and a trityl (Trt) group on the glutamine side-chain amide enabling controlled sequential peptide assembly and minimizing side reactions during synthesis Based on these chemical properties Fmoc-Gln Trt -OH is utilized in biomedical research applications such as peptide drug development protein engineering and studies of peptide structure-function relationships
Catalog No. 50-000-88519