Learn More
Medchemexpress LLC Fmoc-Gln(Trt)-OH | 132327-80-1 | 99.86% | 610.70 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW013081
Encompass_Preferred
Fmoc-Gln(Trt)-OH is a glutamine derivative containing the amine protecting group Fmoc. It can be used in solid-phase peptide synthesis. In peptide synthesis, Fmoc-Gln(Trt)-OH can effectively prevent unnecessary reactions of glutamine during the synthesis process and ensure the correct synthesis of peptide chains.
  • Glutamine derivative containing amine protecting group Fmoc
  • Used in solid-phase peptide synthesis
Catalog No. 50-004-51332