Learn More
Medchemexpress LLC Fmoc-Gln(Trt)-OH | 132327-80-1 | 99.95% | 610.70 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01308110MM/1ML
Encompass_Preferred
Fmoc-Gln(Trt)-OH is a glutamine derivative that contains the amine protecting group Fmoc. It can be utilized in solid-phase peptide synthesis. In peptide synthesis, Fmoc-Gln(Trt)-OH is effective in preventing unnecessary glutamine reactions during the synthesis process, thereby ensuring the accurate synthesis of peptide chains.
  • Glutamine derivative with Fmoc amine protecting group.
  • Applicable in solid-phase peptide synthesis.
  • Prevents undesired reactions of glutamine during synthesis.
  • Ensures correct peptide chain synthesis.
Catalog No. 50-004-54256