Learn More
Apexbio Technology LLC Fmoc-Gln(Trt)-OPfp 132388-65-9 25g
SDP
Supplier:  Apexbio Technology LLC A788225000
Encompass_Preferred
Fmoc-Gln Trt -OPfp is an amino acid derivative utilized in solid-phase peptide synthesis workflows Structurally it features an N-terminal fluorenylmethoxycarbonyl (Fmoc) protective group a trityl (Trt) group on the glutamine side chain and an activated pentafluorophenyl (OPfp) ester at the carboxyl terminus Typical applications include the synthetic assembly of peptides and protein fragments production of bioactive peptide sequences and investigation of peptide structure-function relationships and molecular-level interactions in biomedical research contexts
Catalog No. 50-000-90582