Learn More
Medchemexpress LLC Fmoc-Gln(Trt)-Ser(psi(Me,Me)pro)-OH | 1821378-64-6 | 98.5% | 737.84 | 1 G

Supplier: Medchemexpress LLC HYP23911G
Fmoc-Gln(Trt)-Ser(psi(Me,Me)pro)-OH is a dipeptide.
- Molecular weight: 737.84
- Formula: C45H43N3O7
- CAS number: 1821378-64-6
- Appearance: Solid
- Color: White to off-white
- Purity: 98.5%
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: Store at room temperature for 3 years. In solvent: -80°C for 2 years, -20°C for 1 year.
- Solubility in vitro: H2O
- Target: Amino acid derivatives
- Pathway: Others
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.