Learn More
Sigma Aldrich Fine Chemicals Biosciences fmoc GlnTrt OH Novabiochem
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 8520450250
Encompass
fmoc-Gln(Trt)-OH has good solubility properties in most organic solvents and its use has been shown to result in significantly purer peptides than other derivatives used for the introduction of Gln [1]. Coupling can be performed by standard procedures. The trityl-protecting group is normally removed by 95% TfA in 1-3 hours with no alkylation of Trp.The product number for this product was previously 04-12-1090.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-289-7807