Learn More
Bachem Fmoc-Glu(ODmab)-OH
SDP
Supplier:  Bachem 4027143.0001
Encompass_Preferred
Fmoc-Glu(ODmab)-OH, 268730-86-5, C40H44N2O8, Mr 680.81, 1g. The ODmab group is an excellent protecting group for carboxylic acids. It is orthogonal to both acid (TFA) and 20% piperidine-labile protecting groups used in standard and automated peptide chemistry. Dmab esters are quantitatively cleaved with 2% hydrazine in DMF within minutes.
Catalog No. 50-258-712